siRNA Details
Gene Symbol: HN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002033.2 | NP_001002033.1 | 4, 5 | 686 | ||
NM_001288609.1 | NP_001275538.1 | 3, 4 | 1275 | |||
NM_001288610.1 | NP_001275539.1 | 3, 4 | 638 | |||
NM_001288611.1 | NP_001275540.1 | 3, 4 | 591 | |||
NM_016185.3 | NP_057269.1 | 3, 4 | 591 | |||
GenBank mRNA | AF086910.2 | - | 3, 4 | 591 | ||
AF177862.1 | - | 3, 4 | 408 | |||
AF266846.1 | - | 3, 4 | 420 | |||
AK223321.1 | - | 3, 4 | 420 | |||
AK308253.1 | - | 3, 4 | 638 | |||
AK312613.1 | - | 3, 4 | 420 | |||
AY322170.1 | - | 4, 5 | 678 | |||
BC001420.2 | - | 3, 4 | 409 | 3140086 | ||
BC039343.1 | - | 3, 4 | 1277 | 5269120 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
