siRNA Details
Gene Symbol: CTRB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC005385.1 | - | 7 | 799 | 3950220 | |
BT007356.1 | - | 6 | 757 | |||
M24400.1 | - | 6 | 773 |
Gene Symbol: CTRB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK124403.1 | - | 4 | 2189 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
