siRNA Details
Gene Symbol: SIRPG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039508.1 | NP_001034597.1 | 5 | 1175 | ||
NM_018556.3 | NP_061026.2 | 6 | 1508 | |||
NM_080816.2 | NP_543006.2 | 4 | 857 | |||
XM_005260749.3 | XP_005260806.1 | 5 | 1226 | |||
XM_011529286.2 | XP_011527588.1 | 6 | 1555 | |||
GenBank mRNA | AB042624.1 | - | 6 | 1524 | ||
AK124224.1 | - | 1 | 3888 | |||
AK292745.1 | - | 6 | 1508 | |||
BC020629.1 | - | 4 | 834 | |||
BC064532.1 | - | 6 | 1469 | 5588038 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
