siRNA Details
Gene Symbol: ZNF37A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007094.2 | NP_001007095.1 | 8 | 6887 | ||
NM_001178101.1 | NP_001171572.1 | 7 | 6409 | |||
NM_001324245.1 | NP_001311174.1 | 7 | 6338 | |||
NM_001324246.1 | NP_001311175.1 | 8 | 6883 | |||
NM_001324247.1 | NP_001311176.1 | 8 | 6628 | |||
NM_001324248.1 | NP_001311177.1 | 7 | 6593 | |||
NM_001324249.1 | NP_001311178.1 | 7 | 6589 | |||
NM_001324250.1 | NP_001311179.1 | 8 | 6703 | |||
NM_001324251.1 | NP_001311180.1 | 8 | 6707 | |||
NM_001324259.1 | NP_001311188.1 | 8 | 6937 | |||
NM_001324260.1 | NP_001311189.1 | 7 | 6459 | |||
NM_003421.2 | NP_003412.1 | 7 | 6413 | |||
XM_005252586.3 | XP_005252643.1 | 2 | 6014 | |||
XM_011519656.2 | XP_011517958.1 | 8 | 6795 | |||
XM_011519657.2 | XP_011517959.1 | 8 | 6617 | |||
XM_011519658.2 | XP_011517960.1 | 7 | 6335 | |||
XM_017016618.1 | XP_016872107.1 | 8 | 6932 | |||
XM_017016619.1 | XP_016872108.1 | 7 | 6468 | |||
XM_017016620.1 | XP_016872109.1 | 8 | 6936 | |||
XM_017016622.1 | XP_016872111.1 | 8 | 6932 | |||
GenBank mRNA | AJ492195.1 | - | 7 | 6379 | ||
BX647089.1 | - | 8 | 6873 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
