siRNA Details
Gene Symbol: CCL28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301873.1 | NP_001288802.1 | 3 | 488 | ||
NM_001301874.1 | NP_001288803.1 | 3 | 488 | |||
NM_001301875.1 | NP_001288804.1 | 3 | 579 | |||
NM_148672.3 | NP_683513.1 | 3 | 488 | |||
GenBank mRNA | AF110384.1 | - | 3 | 467 | ||
AF266504.1 | - | 3 | 465 | |||
AK307307.1 | - | 3 | 443 | |||
BC062668.1 | - | 3 | 430 | 30326117 | ||
BC069532.1 | - | 3 | 463 | 7262110 | ||
BX486632.1 | - | 3 | 489 | |||
CD671712.1 | - | 3 | 579 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
