siRNA Details
Gene Symbol: ITPRIPL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008949.2 | NP_001008949.1 | 3 | 1615 | ||
NM_001163523.1 | NP_001156995.1 | 2 | 1505 | |||
NM_001163524.1 | NP_001156996.1 | 2 | 1372 | |||
NM_001324490.1 | NP_001311419.1 | 2 | 1297 | |||
NM_178495.5 | NP_848590.3 | 1 | 1715 | |||
XM_017003427.1 | XP_016858916.1 | 2 | 1548 | |||
GenBank mRNA | BC034503.2 | - | 1 | 1690 | 4824775 | |
BC073153.1 | - | 3 | 1531 | 6018749 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
