siRNA Details
Gene Symbol: ARHGAP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287242.1 | NP_001274171.1 | 9 | 1315 | ||
NM_006125.2 | NP_006116.2 | 9 | 2538 | |||
NM_013423.2 | NP_038267.1 | 9 | 1367 | |||
NM_013427.2 | NP_038286.2 | 9 | 2538 | |||
NR_109776.1 | 10 | 2634 | ||||
GenBank mRNA | AB208792.1 | - | 9 | 1920 | ||
AF012272.2 | - | 9 | 2539 | |||
AF022212.2 | - | 10 | 2635 | |||
AF117067.1 | - | 9 | 2539 | |||
AF177663.1 | - | 9 | 1367 | |||
AF177665.1 | - | 9 | 1173 | |||
AK293078.1 | - | 9 | 1969 | |||
AK297713.1 | - | 9 | 1212 | |||
AK297747.1 | - | 3 | 324 | |||
AK303444.1 | - | 9 | 1228 | |||
BC150635.1 | - | 9 | 1697 | 9057005 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
