siRNA Details
Gene Symbol: ARHGAP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287242.1 | NP_001274171.1 | 6 | 950 | ||
NM_006125.2 | NP_006116.2 | 6 | 2173 | |||
NM_013423.2 | NP_038267.1 | 6 | 1002 | |||
NM_013427.2 | NP_038286.2 | 6 | 2173 | |||
NR_109776.1 | 7 | 2269 | ||||
GenBank mRNA | AB208792.1 | - | 6 | 1555 | ||
AF012272.2 | - | 6 | 2174 | |||
AF022212.2 | - | 7 | 2270 | |||
AF117067.1 | - | 6 | 2174 | |||
AF177663.1 | - | 6 | 1002 | |||
AF177665.1 | - | 6 | 808 | |||
AK293078.1 | - | 6 | 1604 | |||
AK297713.1 | - | 6 | 847 | |||
AK303444.1 | - | 6 | 863 | |||
BC150635.1 | - | 6 | 1332 | 9057005 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
