siRNA Details
Gene Symbol: MAGEB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002363.4 | NP_002354.2 | 4 | 921 | ||
NM_177404.2 | NP_796379.1 | 2 | 764 | |||
NM_177415.2 | NP_803134.1 | 3 | 838 | |||
GenBank mRNA | AK314977.1 | - | 3 | 838 | ||
BC013772.1 | - | 2 | 712 | 3917511 | ||
BU166425.1 | - | 2 | 764 | |||
CR541762.1 | - | 1 | 601 | |||
X82539.1 | - | 4 | 924 |
Gene Symbol: MAGEB4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002367.3 | NP_002358.1 | 1 | 797 | ||
GenBank mRNA | AK313769.1 | - | 1 | 737 | ||
BC032852.2 | - | 1 | 685 | 5267183 | ||
CR541779.1 | - | 1 | 601 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
