siRNA Details
Gene Symbol: SEP15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004261.3 | NP_004252.2 | 5 | 957 | ||
NM_203341.1 | NP_976086.1 | 4 | 907 | |||
GenBank mRNA | AF051894.1 | - | 5 | 639 | ||
AF288991.1 | - | 5 | 639 | |||
AK225252.1 | - | 5 | 648 | |||
AK225640.1 | - | 5 | 647 | |||
AL833575.1 | - | 5 | 660 | |||
BC005294.1 | - | 5 | 665 | 4041321 | ||
BC016359.1 | - | 5 | 652 | 4096759 | ||
BC021697.1 | - | 4 | 621 | 4051151 | ||
BQ429619.1 | - | 5 | 673 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
