siRNA Details
Gene Symbol: SEP15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004261.3 | NP_004252.2 | 5 | 1037 | ||
NM_203341.1 | NP_976086.1 | 4 | 987 | |||
GenBank mRNA | AF051894.1 | - | 5 | 719 | ||
AF288991.1 | - | 5 | 719 | |||
AK225252.1 | - | 5 | 728 | |||
AK225640.1 | - | 5 | 727 | |||
AL833575.1 | - | 5 | 740 | |||
BC005294.1 | - | 5 | 745 | 4041321 | ||
BC016359.1 | - | 5 | 732 | 4096759 | ||
BC021697.1 | - | 4 | 701 | 4051151 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
