siRNA Details
Gene Symbol: RUSC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135999.1 | NP_001129471.1 | 2 | 2122 | ||
NM_014806.3 | NP_055621.1 | 2 | 1866 | |||
NR_052015.2 | 2 | 1866 | ||||
GenBank mRNA | AB002373.2 | - | 2 | 1708 | ||
AK299805.1 | - | 2 | 1812 | |||
BC064843.1 | - | 1 | 910 | |||
BC132766.1 | - | 1 | 1651 | 40146788 | ||
BC132770.1 | - | 1 | 1651 | 40146792 | ||
BC146654.1 | - | 2 | 1840 | 40147826 | ||
BC150211.1 | - | 2 | 1778 | |||
BC150262.1 | - | 2 | 1719 | 9007205 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
