siRNA Details
Gene Symbol: RUSC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135999.1 | NP_001129471.1 | 4 | 3322 | ||
NM_014806.3 | NP_055621.1 | 4 | 3066 | |||
NR_052015.2 | 4 | 2741 | ||||
XM_006716896.2 | XP_006716959.1 | 3 | 2913 | |||
GenBank mRNA | AB002373.2 | - | 4 | 2908 | ||
AK299805.1 | - | 4, 5 | 3012 | |||
BC064843.1 | - | 3 | 2110 | |||
BC132766.1 | - | 3 | 2851 | 40146788 | ||
BC132770.1 | - | 3 | 2851 | 40146792 | ||
BC146654.1 | - | 4 | 3040 | 40147826 | ||
BC150211.1 | - | 4 | 2653 | |||
BC150262.1 | - | 4 | 2919 | 9007205 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
