siRNA Details
Gene Symbol: PHACTR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199505.1 | NP_001186434.1 | 13 | 2038 | ||
NM_001199506.1 | NP_001186435.1 | 13 | 2033 | |||
NM_001281507.1 | NP_001268436.1 | 13 | 2137 | |||
NM_080672.4 | NP_542403.1 | 13 | 2515 | |||
NM_183244.1 | NP_899067.1 | 13 | 2158 | |||
NM_183246.1 | NP_899069.1 | 12 | 1948 | |||
XM_011528525.2 | XP_011526827.1 | 13 | 2144 | |||
XM_011528526.2 | XP_011526828.1 | 12 | 1950 | |||
XM_017027626.1 | XP_016883115.1 | 13 | 2247 | |||
XM_017027627.1 | XP_016883116.1 | 13 | 2192 | |||
XM_017027628.1 | XP_016883117.1 | 12 | 1840 | |||
XM_017027629.1 | XP_016883118.1 | 12 | 1983 | |||
XM_017027630.1 | XP_016883119.1 | 12 | 1811 | |||
XM_017027631.1 | XP_016883120.1 | 13 | 2074 | |||
GenBank mRNA | AB098521.1 | - | 13 | 2158 | ||
AB098522.1 | - | 12 | 1948 | |||
AJ311122.2 | - | 13 | 2514 | |||
AJ617581.1 | - | 13 | 2033 | |||
AK090812.1 | - | 2 | 2194 | |||
AK091632.1 | - | 13 | 2160 | |||
AK098788.1 | - | 13 | 2038 | |||
BC108303.1 | - | 13 | 2515 | 4179728 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
