siRNA Details
Gene Symbol: PHACTR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199505.1 | NP_001186434.1 | 11 | 1458 | ||
NM_001199506.1 | NP_001186435.1 | 11 | 1453 | |||
NM_001281507.1 | NP_001268436.1 | 11 | 1557 | |||
NM_080672.4 | NP_542403.1 | 11 | 1935 | |||
NM_183244.1 | NP_899067.1 | 11 | 1578 | |||
NM_183246.1 | NP_899069.1 | 10 | 1368 | |||
XM_011528525.2 | XP_011526827.1 | 11 | 1564 | |||
XM_011528526.2 | XP_011526828.1 | 10 | 1370 | |||
XM_017027626.1 | XP_016883115.1 | 11 | 1667 | |||
XM_017027627.1 | XP_016883116.1 | 11 | 1612 | |||
XM_017027628.1 | XP_016883117.1 | 10 | 1260 | |||
XM_017027629.1 | XP_016883118.1 | 10 | 1403 | |||
XM_017027630.1 | XP_016883119.1 | 10 | 1231 | |||
XM_017027631.1 | XP_016883120.1 | 11 | 1494 | |||
GenBank mRNA | AB098521.1 | - | 11 | 1578 | ||
AB098522.1 | - | 10 | 1368 | |||
AJ311122.2 | - | 11 | 1934 | |||
AJ617581.1 | - | 11 | 1453 | |||
AK091632.1 | - | 11 | 1580 | |||
AK098788.1 | - | 11 | 1458 | |||
AK295459.1 | - | 11 | 1557 | |||
AK314493.1 | - | 11 | 1665 | |||
AK316047.1 | - | 11 | 1450 | |||
BC108303.1 | - | 11 | 1935 | 4179728 | ||
BC117362.1 | - | 11 | 1823 | 40125913 | ||
BC117364.1 | - | 11 | 1826 | 40125915 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
