siRNA Details
Gene Symbol: LDLRAD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181481.4 | NP_852146.1 | 2, 3 | 512 | ||
NM_181482.4 | NP_852147.1 | 3, 4 | 771 | |||
XM_005258140.1 | XP_005258197.1 | 3, 4 | 741 | |||
XM_006722353.1 | XP_006722416.1 | 4, 5 | 798 | |||
XM_006722354.1 | XP_006722417.1 | 2, 3 | 602 | |||
GenBank mRNA | AF009424.1 | - | 2, 3 | 515 | ||
AF009425.1 | - | 2, 3 | 515 | |||
BC030199.1 | - | 3, 4 | 705 | 5199354 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
