siRNA Details
Gene Symbol: EIF1AY | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278612.1 | NP_001265541.1 | 6 | 1185 | ||
NM_004681.3 | NP_004672.2 | 7 | 1236 | |||
GenBank mRNA | AF000987.1 | - | 7 | 1207 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
