siRNA Details
Gene Symbol: DZIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014934.4 | NP_055749.1 | 22 | 3800 | ||
NM_198968.3 | NP_945319.1 | 23 | 3857 | |||
XM_006719917.2 | XP_006719980.1 | 22 | 3995 | |||
XM_017020444.1 | XP_016875933.1 | 21 | 3935 | |||
XM_017020446.1 | XP_016875935.1 | 24 | 3957 | |||
GenBank mRNA | AB023213.1 | - | 22 | 3783 | ||
AK094042.1 | - | 9 | 1821 | |||
AK094990.1 | - | 1 | 1686 | |||
AL049931.1 | - | 6 | 1161 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
