siRNA Details
Gene Symbol: DZIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014934.4 | NP_055749.1 | 22 | 5039 | ||
NM_198968.3 | NP_945319.1 | 23 | 5096 | |||
GenBank mRNA | AK092363.1 | - | 1 | 389 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
