siRNA Details
Gene Symbol: DZIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014934.4 | NP_055749.1 | 9 | 1888 | ||
NM_198968.3 | NP_945319.1 | 9 | 1888 | |||
XM_005254032.1 | XP_005254089.1 | 8 | 2026 | |||
XM_006719916.1 | XP_006719979.1 | 8 | 2026 | |||
XM_006719917.2 | XP_006719980.1 | 8 | 2026 | |||
XM_006719918.1 | XP_006719981.1 | 8 | 2026 | |||
XM_006719920.1 | XP_006719983.1 | 9 | 1888 | |||
XM_017020444.1 | XP_016875933.1 | 8 | 2026 | |||
XM_017020445.1 | XP_016875934.1 | 10 | 1988 | |||
XM_017020446.1 | XP_016875935.1 | 10 | 1988 | |||
XM_017020447.1 | XP_016875936.1 | 5 | 524 | |||
GenBank mRNA | AB023213.1 | - | 9 | 1871 | ||
AF272347.1 | - | 6 | 1033 | |||
AF272348.2 | - | 6 | 1319 | |||
BC041804.1 | - | 9 | 1888 | 5271595 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
