siRNA Details
Gene Symbol: KIAA0753 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014804.2 | NP_055619.2 | 19 | 4299 | ||
XM_006721612.2 | XP_006721675.1 | 17 | 3742 | |||
XM_011524090.2 | XP_011522392.1 | 20 | 4365 | |||
XM_011524091.2 | XP_011522393.1 | 20 | 5750 | |||
XM_011524095.2 | XP_011522397.1 | 20 | 4097 | |||
XM_011524098.2 | XP_011522400.1 | 18 | 3996 | |||
XM_011524099.2 | XP_011522401.1 | 18 | 3486 | |||
XM_011524100.2 | XP_011522402.1 | 17 | 3268 | |||
XM_017025455.1 | XP_016880944.1 | 20 | 4308 | |||
XM_017025456.1 | XP_016880945.1 | 17 | 4060 | |||
XM_017025457.1 | XP_016880946.1 | 16 | 3830 | |||
GenBank mRNA | AB018296.1 | - | 19 | 4044 | ||
AL080108.1 | - | 13 | 2732 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
