siRNA Details
Gene Symbol: KIAA0753 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014804.2 | NP_055619.2 | 19 | 3472 | ||
XM_006721612.2 | XP_006721675.1 | 17 | 2915 | |||
XM_011524090.2 | XP_011522392.1 | 20 | 3538 | |||
XM_011524091.2 | XP_011522393.1 | 20 | 4923 | |||
XM_011524095.2 | XP_011522397.1 | 20 | 3270 | |||
XM_011524098.2 | XP_011522400.1 | 18 | 3169 | |||
XM_011524099.2 | XP_011522401.1 | 18 | 2659 | |||
XM_011524100.2 | XP_011522402.1 | 17 | 2441 | |||
XM_017025455.1 | XP_016880944.1 | 20 | 3481 | |||
XM_017025456.1 | XP_016880945.1 | 17 | 3233 | |||
XM_017025457.1 | XP_016880946.1 | 16 | 3003 | |||
GenBank mRNA | AB018296.1 | - | 19 | 3217 | ||
AK292876.1 | - | 19 | 3234 | |||
AK296971.1 | - | 18 | 3114 | |||
AL080108.1 | - | 13 | 1905 | |||
BC113016.1 | - | 18 | 3187 | 40036441 | ||
BC113017.1 | - | 17 | 3080 | 40036446 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
