siRNA Details
Gene Symbol: KIAA0753 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014804.2 | NP_055619.2 | 9 | 1948 | ||
XM_006721612.2 | XP_006721675.1 | 7 | 1391 | |||
XM_011524090.2 | XP_011522392.1 | 9 | 1948 | |||
XM_011524091.2 | XP_011522393.1 | 9 | 3333 | |||
XM_011524095.2 | XP_011522397.1 | 9 | 1680 | |||
XM_011524096.1 | XP_011522398.1 | 9 | 1948 | |||
XM_011524098.2 | XP_011522400.1 | 7 | 1579 | |||
XM_011524099.2 | XP_011522401.1 | 7 | 1069 | |||
XM_011524100.2 | XP_011522402.1 | 6 | 851 | |||
XM_017025455.1 | XP_016880944.1 | 9 | 1891 | |||
XM_017025456.1 | XP_016880945.1 | 7 | 1709 | |||
XM_017025457.1 | XP_016880946.1 | 6 | 1479 | |||
GenBank mRNA | AB018296.1 | - | 9 | 1693 | ||
AK292876.1 | - | 9 | 1710 | |||
AK296971.1 | - | 8 | 1590 | |||
AK300547.1 | - | 2 | 96 | |||
AL080108.1 | - | 3 | 381 | |||
BC017566.1 | - | 9 | 1707 | |||
BC113016.1 | - | 8 | 1663 | 40036441 | ||
BC113017.1 | - | 7 | 1556 | 40036446 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
