siRNA Details
Gene Symbol: SP8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182700.5 | NP_874359.2 | 2 | 987 | ||
NM_198956.3 | NP_945194.1 | 3 | 1083 | |||
GenBank mRNA | AK056857.1 | - | 3 | 816 | ||
AK290465.1 | - | 3 | 861 | |||
AK299947.1 | - | 3 | 758 | |||
AY167047.1 | - | 2 | 984 | |||
AY167048.1 | - | 2 | 972 | |||
BC038669.1 | - | 3 | 858 | 5259444 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
