siRNA Details
Gene Symbol: FCRL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159488.1 | NP_001152960.1 | 7 | 1324 | ||
NM_030764.3 | NP_110391.2 | 7 | 1324 | |||
NR_125358.1 | 4 | 472 | ||||
XM_011509974.2 | XP_011508276.1 | 7 | 1659 | |||
XM_011509975.2 | XP_011508277.1 | 7 | 1661 | |||
XM_011509976.2 | XP_011508278.1 | 7 | 1660 | |||
XM_017002316.1 | XP_016857805.1 | 7 | 1659 | |||
XM_017002317.1 | XP_016857806.1 | 7 | 1462 | |||
XM_017002318.1 | XP_016857807.1 | 6 | 1382 | |||
XM_017002319.1 | XP_016857808.1 | 6 | 1374 | |||
GenBank mRNA | AF319438.1 | - | 2 | 809 | ||
AF319439.1 | - | 2 | 647 | |||
AF390037.1 | - | 2 | 647 | |||
AF459633.1 | - | 7 | 1312 | |||
AK301113.1 | - | 7 | 1311 | |||
AL833194.1 | - | 2 | 3183 | |||
AY043465.1 | - | 7 | 1311 | |||
AY358130.1 | - | 7 | 1314 | |||
BC069185.1 | - | 4 | 476 | |||
BC129836.1 | - | 2 | 649 | 40115446 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
