siRNA Details
Gene Symbol: MUC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002457.3 | NP_002448.3 | 15304 | |||
GenBank mRNA | L21998.1 | - | 15304 | |||
M94132.1 | - | 19 | 2692 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
