siRNA Details
Gene Symbol: WNT8A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300939.1 | NP_001287868.1 | 5 | 1285 | ||
NM_058244.3 | NP_490645.1 | 6 | 1193 | |||
NR_125351.1 | 6 | 1193 | ||||
XM_017009825.1 | XP_016865314.1 | 6 | 1263 | |||
XM_017009826.1 | XP_016865315.1 | 7 | 1125 | |||
GenBank mRNA | AB057725.1 | - | 6 | 1000 | ||
AY009402.1 | - | 6 | 1095 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
