siRNA Details
Gene Symbol: A1CF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198818.1 | NP_001185747.1 | 8 | 964 | ||
NM_001198819.1 | NP_001185748.1 | 9 | 1107 | |||
NM_001198820.1 | NP_001185749.1 | 8 | 1014 | |||
NM_014576.3 | NP_055391.2 | 7 | 871 | |||
NM_138932.2 | NP_620310.1 | 7 | 871 | |||
NM_138933.2 | NP_620311.1 | 7 | 966 | |||
XM_005269718.2 | XP_005269775.1 | 8 | 916 | |||
XM_005269720.3 | XP_005269777.1 | 6 | 829 | |||
XM_011539729.2 | XP_011538031.1 | 6 | 808 | |||
XM_017016160.1 | XP_016871649.1 | 6 | 829 | |||
GenBank mRNA | AF209192.1 | - | 7 | 823 | ||
AF271789.1 | - | 5 | 683 | |||
AF271790.1 | - | 6 | 833 | |||
AJ272078.1 | - | 5 | 687 | |||
AJ272079.1 | - | 5 | 687 | |||
AK000324.1 | - | 7 | 965 | |||
AK291982.1 | - | 8 | 916 | |||
AK303801.1 | - | 9 | 1061 | |||
BC130519.1 | - | 6 | 768 | 40146550 | ||
BC144196.1 | - | 7 | 911 | 9052716 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
