siRNA Details
Gene Symbol: A1CF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198818.1 | NP_001185747.1 | 7 | 844 | ||
NM_001198819.1 | NP_001185748.1 | 8 | 987 | |||
NM_001198820.1 | NP_001185749.1 | 7 | 894 | |||
NM_014576.3 | NP_055391.2 | 6 | 751 | |||
NM_138932.2 | NP_620310.1 | 6 | 751 | |||
NM_138933.2 | NP_620311.1 | 6 | 846 | |||
XM_005269718.2 | XP_005269775.1 | 7 | 796 | |||
XM_005269720.3 | XP_005269777.1 | 5 | 709 | |||
XM_011539729.2 | XP_011538031.1 | 5 | 688 | |||
XM_011539730.2 | XP_011538032.1 | 8 | 939 | |||
XM_017016160.1 | XP_016871649.1 | 5 | 709 | |||
GenBank mRNA | AF271789.1 | - | 4 | 563 | ||
AF271790.1 | - | 5 | 713 | |||
AJ272078.1 | - | 4 | 567 | |||
AJ272079.1 | - | 4 | 567 | |||
AK000324.1 | - | 6 | 845 | |||
AK291982.1 | - | 7 | 796 | |||
AK303801.1 | - | 8 | 941 | |||
BC130519.1 | - | 5 | 648 | 40146550 | ||
BC144196.1 | - | 6 | 791 | 9052716 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
