siRNA Details
Gene Symbol: PLP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000533.4 | NP_000524.3 | 3 | 510 | ||
NM_001128834.2 | NP_001122306.1 | 4 | 592 | |||
NM_001305004.1 | NP_001291933.1 | 3 | 285 | |||
NM_199478.2 | NP_955772.1 | 3 | 510 | |||
GenBank mRNA | AK128782.1 | - | 3 | 433 | ||
AK292728.1 | - | 4 | 592 | |||
AK295388.1 | - | 3 | 264 | |||
AK308966.1 | - | 2 | 239 | |||
AK309239.1 | - | 3 | 451 | |||
AK312340.1 | - | 3 | 433 | |||
BC002665.1 | - | 3 | 453 | 3606269 | ||
BC095452.1 | - | 3 | 394 | 30717624 | ||
BQ717999.1 | - | 3 | 408 | |||
BT019601.1 | - | 2 | 307 | |||
BT019602.1 | - | 2 | 307 | |||
BX445448.2 | - | 3 | 411 | |||
CA389667.1 | - | 3 | 495 | |||
CR536542.1 | - | 2 | 307 | |||
DA299940.1 | - | 3 | 511 | |||
KU178261.1 | - | 2 | 307 | |||
KU178262.1 | - | 2 | 307 | |||
M17085.1 | - | 2 | 307 | |||
M27110.1 | - | 3 | 425 | |||
M54927.1 | - | 3 | 428 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
