siRNA Details
Gene Symbol: PLP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000533.4 | NP_000524.3 | 7 | 2320 | ||
NM_001128834.2 | NP_001122306.1 | 8 | 2402 | |||
NM_001305004.1 | NP_001291933.1 | 7 | 2095 | |||
NM_199478.2 | NP_955772.1 | 7 | 2215 | |||
GenBank mRNA | AK128782.1 | - | 7 | 2243 | ||
BC002665.1 | - | 7 | 2158 | 3606269 | ||
BC095452.1 | - | 7 | 2204 | 30717624 | ||
EB387051.1 | - | 1 | 560 | |||
M27110.1 | - | 7 | 2238 | |||
M54927.1 | - | 7 | 2235 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
