siRNA Details
Gene Symbol: DIEXF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014388.6 | NP_055203.4 | 12 | 7997 | ||
XM_006711275.3 | XP_006711338.1 | 10 | 7638 | |||
GenBank mRNA | AK022766.1 | - | 1 | 2024 | ||
CR749825.1 | - | 12 | 7932 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
