siRNA Details
Gene Symbol: CALD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004342.6 | NP_004333.1 | 6 | 1148 | ||
NM_033138.3 | NP_149129.2 | 7 | 1913 | |||
NM_033139.3 | NP_149130.1 | 5 | 963 | |||
NM_033140.3 | NP_149131.1 | 4 | 885 | |||
NM_033157.3 | NP_149347.2 | 7 | 1226 | |||
XM_017012650.1 | XP_016868139.1 | 5 | 1448 | |||
XM_017012651.1 | XP_016868140.1 | 5 | 761 | |||
XM_017012652.1 | XP_016868141.1 | 5 | 922 | |||
XM_017012653.1 | XP_016868142.1 | 4 | 683 | |||
XM_017012654.1 | XP_016868143.1 | 4 | 885 | |||
GenBank mRNA | AF247820.3 | - | 6 | 911 | ||
AK289686.1 | - | 7 | 1219 | |||
AK298524.1 | - | 6 | 774 | |||
AK300282.1 | - | 4 | 854 | |||
AK304729.1 | - | 6 | 695 | |||
BC014035.2 | - | 4 | 848 | |||
BC040354.1 | - | 6 | 883 | 4753285 | ||
BX648808.1 | - | 4 | 846 | |||
D90452.1 | - | 5 | 753 | |||
D90453.1 | - | 4 | 675 | |||
M64110.1 | - | 5 | 793 | |||
M83216.1 | - | 7 | 1676 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
