siRNA Details
Gene Symbol: CALD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004342.6 | NP_004333.1 | 14 | 2973 | ||
NM_033138.3 | NP_149129.2 | 15 | 3738 | |||
NM_033139.3 | NP_149130.1 | 13 | 2788 | |||
NM_033140.3 | NP_149131.1 | 12 | 2710 | |||
NM_033157.3 | NP_149347.2 | 15 | 3048 | |||
XM_017012650.1 | XP_016868139.1 | 13 | 3270 | |||
XM_017012651.1 | XP_016868140.1 | 13 | 2586 | |||
XM_017012652.1 | XP_016868141.1 | 13 | 2747 | |||
XM_017012653.1 | XP_016868142.1 | 12 | 2505 | |||
XM_017012654.1 | XP_016868143.1 | 12 | 2707 | |||
GenBank mRNA | AU134367.1 | - | 1 | 312 | ||
BC040354.1 | - | 14 | 2709 | 4753285 | ||
BX538339.1 | - | 4 | 1213 | |||
BX648808.1 | - | 12 | 2667 | |||
BX649088.1 | - | 5 | 1267 | |||
M64110.1 | - | 13 | 2620 | |||
M83216.1 | - | 15 | 3499 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
