siRNA Details
Gene Symbol: CALD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004342.6 | NP_004333.1 | 14 | 3244 | ||
NM_033138.3 | NP_149129.2 | 15 | 4009 | |||
NM_033139.3 | NP_149130.1 | 13 | 3059 | |||
NM_033140.3 | NP_149131.1 | 12 | 2981 | |||
NM_033157.3 | NP_149347.2 | 15 | 3319 | |||
XM_017012650.1 | XP_016868139.1 | 13 | 3541 | |||
XM_017012651.1 | XP_016868140.1 | 13 | 2857 | |||
XM_017012652.1 | XP_016868141.1 | 13 | 3018 | |||
XM_017012653.1 | XP_016868142.1 | 12 | 2776 | |||
XM_017012654.1 | XP_016868143.1 | 12 | 2978 | |||
GenBank mRNA | AU134367.1 | - | 1 | 583 | ||
BC040354.1 | - | 14 | 2980 | 4753285 | ||
BX538339.1 | - | 4 | 1484 | |||
BX648808.1 | - | 12 | 2938 | |||
BX649088.1 | - | 5 | 1538 | |||
CV573441.1 | - | 1 | 18 | |||
M64110.1 | - | 13 | 2891 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
