siRNA Details
Gene Symbol: TBKBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014726.2 | NP_055541.1 | 9 | 3062 | ||
XM_005257859.4 | XP_005257916.1 | 10 | 2361 | |||
XM_005257860.4 | XP_005257917.1 | 10 | 3186 | |||
XM_005257861.4 | XP_005257918.1 | 10 | 2666 | |||
XM_005257862.3 | XP_005257919.1 | 10 | 2286 | |||
XM_017025431.1 | XP_016880920.1 | 9 | 3567 | |||
XM_017025432.1 | XP_016880921.1 | 9 | 3566 | |||
GenBank mRNA | AB018318.1 | - | 9 | 3062 | ||
BC111418.1 | - | 6 | 1511 | |||
BC152407.1 | - | 9 | 3062 | 8862510 | ||
BC167150.1 | - | 9 | 3062 | 8862522 | ||
DT217050.1 | - | 1 | 289 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
