siRNA Details
Gene Symbol: TBKBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014726.2 | NP_055541.1 | 4 | 1438 | ||
XM_005257859.4 | XP_005257916.1 | 5 | 737 | |||
XM_005257860.4 | XP_005257917.1 | 5 | 1562 | |||
XM_005257861.4 | XP_005257918.1 | 5 | 1042 | |||
XM_005257862.3 | XP_005257919.1 | 5 | 662 | |||
XM_017025431.1 | XP_016880920.1 | 4 | 1946 | |||
XM_017025432.1 | XP_016880921.1 | 4 | 1942 | |||
GenBank mRNA | AB018318.1 | - | 4 | 1438 | ||
BC152407.1 | - | 4 | 1438 | 8862510 | ||
BC167150.1 | - | 4 | 1438 | 8862522 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
