siRNA Details
Gene Symbol: TBKBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014726.2 | NP_055541.1 | 6, 7 | 1724 | ||
XM_005257859.4 | XP_005257916.1 | 7, 8 | 1023 | |||
XM_005257860.4 | XP_005257917.1 | 7, 8 | 1848 | |||
XM_005257861.4 | XP_005257918.1 | 7, 8 | 1328 | |||
XM_005257862.3 | XP_005257919.1 | 7, 8 | 948 | |||
XM_017025431.1 | XP_016880920.1 | 6, 7 | 2229 | |||
XM_017025432.1 | XP_016880921.1 | 6, 7 | 2228 | |||
GenBank mRNA | AB018318.1 | - | 6, 7 | 1724 | ||
BC152407.1 | - | 6, 7 | 1724 | 8862510 | ||
BC167150.1 | - | 6, 7 | 1724 | 8862522 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
