siRNA Details
Gene Symbol: SBNO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100122.1 | NP_001093592.1 | 7 | 843 | ||
NM_014963.2 | NP_055778.2 | 10 | 1175 | |||
XM_011527804.2 | XP_011526106.1 | 10 | 1148 | |||
GenBank mRNA | AB023180.1 | - | 10 | 1152 | ||
AK074102.1 | - | 6 | 635 | |||
AK125139.1 | - | 7 | 790 | |||
AK292407.1 | - | 10 | 1175 | |||
AK296849.1 | - | 7 | 790 | |||
AK300981.1 | - | 11 | 1115 | |||
AK309167.1 | - | 7 | 790 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
