siRNA Details
Gene Symbol: SBNO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100122.1 | NP_001093592.1 | 23 | 2893 | ||
NM_014963.2 | NP_055778.2 | 26 | 3225 | |||
XM_011527804.2 | XP_011526106.1 | 26 | 3198 | |||
GenBank mRNA | AB023180.1 | - | 26 | 3202 | ||
AK074102.1 | - | 22 | 2685 | |||
AK125139.1 | - | 23 | 2840 | |||
AK292407.1 | - | 26 | 3225 | |||
BC106021.1 | - | 3 | 297 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
