siRNA Details
Gene Symbol: L3MBTL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007102.2 | NP_001007103.1 | 22 | 2640 | ||
NM_032438.2 | NP_115814.1 | 23 | 2715 | |||
XM_005267161.4 | XP_005267218.1 | 23 | 2891 | |||
XM_006715576.3 | XP_006715639.1 | 23 | 2820 | |||
XM_006715578.3 | XP_006715641.1 | 23 | 2699 | |||
XM_011536179.2 | XP_011534481.1 | 23 | 2698 | |||
XM_011536180.2 | XP_011534482.1 | 23 | 2717 | |||
XM_011536181.2 | XP_011534483.1 | 23 | 2652 | |||
XM_011536183.2 | XP_011534485.1 | 23 | 3248 | |||
XM_011536184.2 | XP_011534486.1 | 23 | 2652 | |||
XM_017011355.1 | XP_016866844.1 | 22 | 2731 | |||
XM_017011356.1 | XP_016866845.1 | 22 | 2623 | |||
GenBank mRNA | AB058701.1 | - | 22 | 2613 | ||
AK092756.1 | - | 13 | 1892 | |||
AK094137.1 | - | 23 | 2722 | |||
BC060845.1 | - | 22 | 2634 | 30342272 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
