siRNA Details
Gene Symbol: L3MBTL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007102.2 | NP_001007103.1 | 3 | 264 | ||
NM_032438.2 | NP_115814.1 | 3 | 264 | |||
XM_005267161.4 | XP_005267218.1 | 3 | 440 | |||
XM_006715576.3 | XP_006715639.1 | 3 | 264 | |||
XM_006715578.3 | XP_006715641.1 | 3 | 248 | |||
XM_011536179.2 | XP_011534481.1 | 3 | 247 | |||
XM_011536180.2 | XP_011534482.1 | 3 | 266 | |||
XM_011536181.2 | XP_011534483.1 | 3 | 201 | |||
XM_011536183.2 | XP_011534485.1 | 3 | 797 | |||
XM_011536184.2 | XP_011534486.1 | 3 | 201 | |||
XM_017011355.1 | XP_016866844.1 | 3 | 355 | |||
XM_017011356.1 | XP_016866845.1 | 3 | 247 | |||
GenBank mRNA | AB058701.1 | - | 2 | 162 | ||
AK094137.1 | - | 3 | 270 | |||
AK290219.1 | - | 3 | 256 | |||
BC060845.1 | - | 3 | 258 | 30342272 | ||
DA927626.1 | - | 3 | 264 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
