siRNA Details
Gene Symbol: LINC00310 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027267.1 | 2 | 176 | |||
GenBank mRNA | AF426258.1 | - | 2 | 176 | ||
BC093749.1 | - | 2 | 106 | |||
BC111996.1 | - | 2 | 106 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
