siRNA Details
Gene Symbol: INPP5F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014937.3 | NP_055752.1 | 13 | 1717 | ||
XM_006717720.3 | XP_006717783.1 | 13 | 4104 | |||
XM_011539524.2 | XP_011537826.1 | 13 | 1480 | |||
XM_011539525.2 | XP_011537827.1 | 14 | 4187 | |||
XM_011539527.2 | XP_011537829.2 | 12 | 1482 | |||
XM_011539528.2 | XP_011537830.1 | 12 | 1604 | |||
XM_011539529.1 | XP_011537831.1 | 2 | 138 | |||
XM_017015935.1 | XP_016871424.1 | 13 | 1471 | |||
XM_017015936.1 | XP_016871425.1 | 16 | 2105 | |||
GenBank mRNA | AB023183.1 | - | 13 | 1667 | ||
AL137528.1 | - | 7 | 682 | |||
BC111493.1 | - | 13 | 1628 | 6166990 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
