siRNA Details
Gene Symbol: INPP5F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243194.1 | NP_001230123.1 | 6 | 1027 | ||
NM_014937.3 | NP_055752.1 | 20 | 2846 | |||
XM_011539524.2 | XP_011537826.1 | 20 | 2609 | |||
XM_011539525.2 | XP_011537827.1 | 21 | 5316 | |||
XM_011539527.2 | XP_011537829.2 | 19 | 2611 | |||
XM_011539528.2 | XP_011537830.1 | 19 | 2733 | |||
XM_011539529.1 | XP_011537831.1 | 9 | 1267 | |||
XM_017015935.1 | XP_016871424.1 | 20 | 2600 | |||
XM_017015936.1 | XP_016871425.1 | 23 | 3234 | |||
GenBank mRNA | AB023183.1 | - | 20 | 2796 | ||
AK091448.1 | - | 6 | 1567 | |||
AK123633.1 | - | 6 | 1009 | |||
AL137528.1 | - | 14 | 1811 | |||
BC111493.1 | - | 20 | 2757 | 6166990 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
