siRNA Details
Gene Symbol: MS4A7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021201.4 | NP_067024.1 | 7 | 2932 | ||
NM_206938.1 | NP_996821.1 | 6 | 2797 | |||
NM_206939.1 | NP_996822.1 | 7 | 2884 | |||
NM_206940.1 | NP_996823.1 | 6 | 2749 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
