siRNA Details
Gene Symbol: MS4A7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021201.4 | NP_067024.1 | 7 | 1432 | ||
NM_206938.1 | NP_996821.1 | 6 | 1297 | |||
NM_206939.1 | NP_996822.1 | 7 | 1384 | |||
NM_206940.1 | NP_996823.1 | 6 | 1249 | |||
GenBank mRNA | AK075106.1 | - | 7 | 1436 | ||
AK297027.1 | - | 6 | 1233 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
