siRNA Details
Gene Symbol: MAG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199216.1 | NP_001186145.1 | 4 | 361 | ||
NM_002361.3 | NP_002352.1 | 4 | 374 | |||
NM_080600.2 | NP_542167.1 | 4 | 374 | |||
GenBank mRNA | AK222649.1 | - | 4 | 299 | ||
AK222680.1 | - | 4 | 277 | |||
AK223562.1 | - | 4 | 335 | |||
AK294644.1 | - | 4 | 268 | |||
AK308976.1 | - | 4 | 286 | |||
BC053347.1 | - | 4 | 266 | 5177296 | ||
BC093045.1 | - | 4 | 326 | 30396005 | ||
BI755065.1 | - | 4 | 324 | |||
DC331168.1 | - | 4 | 374 | |||
M29273.1 | - | 4 | 296 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
