siRNA Details
Gene Symbol: MAG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199216.1 | NP_001186145.1 | 11 | 2337 | ||
NM_002361.3 | NP_002352.1 | 11 | 2350 | |||
NM_080600.2 | NP_542167.1 | 12 | 2395 | |||
GenBank mRNA | AK094545.1 | - | 12 | 2196 | ||
AK222649.1 | - | 11 | 2275 | |||
AK222680.1 | - | 11 | 2253 | |||
AK223562.1 | - | 11 | 2311 | |||
AK294644.1 | - | 11 | 2244 | |||
BC053347.1 | - | 11 | 2242 | 5177296 | ||
BC093045.1 | - | 12 | 2347 | 30396005 | ||
M29273.1 | - | 11 | 2271 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
