siRNA Details
Gene Symbol: STH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007532.2 | NP_001007533.1 | 1 | 197 | ||
GenBank mRNA | AY179170.1 | - | 1 | 167 | ||
BC130319.1 | - | 1 | 197 | 40146350 | ||
BC130321.1 | - | 1 | 197 | 40146352 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
